Drucken
Magnetrührer, MST und MICROSTIRRER
Lieferant: VELP SCIENTIFIC
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
F203A0161
442-1092EA 110 EUR
442-1092
Magnetrührer, MST und MICROSTIRRER
Rührer Magnetrührer
Kleine, einfache und effektive Rührer speziell für Mikrotitrationen.
  • Stirrers stay cold even after several days of continuous use
  • Particularly appreciated in microbiology and biochemistry
  • Chemically resistant housing
  • Excellent speed control even at low revolutions
  • Epoxy-coated tough steel casing
Pale surface for visualisation of particles and color changes. Remains cold, even after days of continuous use.
Electronic speed regulation: Up to 1100 min⁻¹
Bestellinformation: Designed for the longest lifespan and guaranteed with two years warranty.