Drucken
VWR®, Support Stand, Rectangular Base, Cast Iron
Lieferant: VWR Collection
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
444-0918EA 180 EUR
444-0918
VWR®, Support Stand, Rectangular Base, Cast Iron
Stative
Cast iron support stand bases are constructed with black enamel finish for chemical resistance and durability.
It features a built-in support rod holder with locking knob that accepts a 13 mm diameter support rod.
Bestellinformation: Support rods are not included.