Drucken
Bottle/tube rollers
Lieferant: BARNSTEAD THERMOLYNE
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
88881004
444-0960EA 1290 EUR
444-0960
Bottle/tube rollers
Schüttler Rotations-/Wippschüttler
This sleek compact design unit is capable of rolling multiple size tubes or bottles for applications like cell culture, hybridisation assays and liquid mixing. The anti-corrosive, wear-resistant construction enables use in humid and CO₂ environments.
  • Adjustable speed accommodates a variety of applications
  • Adjustable rollers accommodate a variety of tube/bottle sizes
  • Stacking kit allows two units to be stacked, saving valuable bench space
  • Optional remote control facilitates operating unit while in incubator and adds timer function
Zertifizierungen: cCSAus compliance.
Lieferumfang: Includes O-rings to ensure smooth rolling when using tubes. Bars must be ordered seperately.