Drucken
Precision hotplate, PZ
Lieferant: GESTIGKEIT HARRY
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
PZ28-1 PZ28-2 PZ35 PZ60 PZ72
453-2131EA 1630 EUR
453-2131 453-2132 453-2133 453-2134 453-2135
Precision hotplate, PZ
Heizgeräte Heizplatten
Precision hotplates are equipped with adjustable temperature sensors +5 to +300 °C (PZ28-1 +30 to +110 °C). These sensors function as an emergency control which protect both the appliance and the substance to be heated against dangerous overheating.