Phosphate-Buffered saline (PBS), 10X concentrate, pH 7,4, molecular biology grade, Corning®
Lieferant: MEDIATECHGesamtbewertung: 0
Bewertung: von 5
Phosphate Buffered Saline (PBS) is used to maintain a physiological pH and osmolarity, two key factors in tissue culture and molecular biology research
- Manufactured in a cGMP environment
- Formulated with high quality WFI water
- RNase -, DNase-, and Protease-free
- PBS without calcium and magnesium
- pH 7.4 ±0.1
Phosphate Buffered Saline is used to maintain a physiological pH and osmolarity, two key factors in tissue culture and molecular biology research. This concentrated form should be diluted to a working concentration using reagent grade water.
Achtung! Bitte beachten: For research use only.