Drucken
Heat transfer liquids, therm 250
Lieferant: LAUDA
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
LZB322 LZB122 LZB222
461-2908EA 4210 EUR
461-2908 461-2120 461-2907
Heat transfer liquids, therm 250
Flüssigkeiten für Umwälzbäder
These transfer fluids allow highly accurate thermostating, even at extreme temperatures.
  • Durability
  • Simple and safe handling
  • Reliability, suitable for long-term operation
Recommended temperature range: 50 to 250 °C (open system).
* Denotes silicone oil