Drucken
Wärme- und Umluftschrank WU 6100
Lieferant: HERAEUS
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
51005691
466-2310EA 18500 EUR
466-2310
Wärme- und Umluftschrank WU 6100
Öfen Standard-Trockenschränke
This forced-air circulation oven is ideal for high quality materials testing. For exacting heat processes up to 500 °C.
  • Thermicon P electronic controller with timer, 9 segments
  • Provides precise and even temperature distribution and selected temperature is accurrately maintained
  • Air outlet pipe with vent
  • Excellent thermal insulation ensuring low energy consumption and surface temperatures without risk of burns (40 to 60 °C)
Safety protection class 2 (DIN 12880, part 1).
Various options are available.