Drucken
Desiccator cabinets, small stacking, dry-keeper™, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
H420530000
468-4101EA 666 EUR
468-4101
Desiccator cabinets, small stacking, dry-keeper™, SP Bel-Art
Exsikkatoren Trockenkammern
Small, clear polystyrene desiccator cabinet, these can be safely stacked up to five units high for maximum use of dry storage space.
Lieferumfang: Supplied with ready to use jar of silica gel.