Drucken
Cooling boxes, Icetime® Plus
Lieferant: CAMPING GAZ
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
2000024966
479-2176EA 0 EUR
479-2176
Cooling boxes, Icetime® Plus
Boxen Kühl- und Gefrierboxen, Kühl- und Gefrierbeutel
These thick, high-quality polyurethane insulated extreme coolers, with the new antimicrobial liner keeps the contents fresh and protected from odour, mold and mildew.
  • Tough outer shell and has two cup-holders
  • Premium PU insulated lid for even better performance
  • Suitable for longer camping trips or outings
  • Bail handle for comfortable carrying
Antimicrobial effect is obtained from an additive which is melted down into the plastic before it is molded into the liner.