Medium, Ham's F-12, Corning®
Lieferant: MEDIATECHGesamtbewertung: 0
Bewertung: von 5
Ham's F-12 media was originally formulated for the serum-free culture of CHO cells, and is based on Ham's F-10 medium with increased concentrations of choline, inositol, putrescine, and several amino acids.
- Low endotoxin - < 0,25 EU/ml
- Mycoplasma tested
- Liquid media formulations are formulated with cell culture grade WFI water
- Manufactured in a cGMP environment
This medium is also suitable for carcinoma cells, rat skeletal myoblasts, Chinese hamster lung cells, and rat, rabbit, and chicken embryos.