Drucken

Anti-CD45R Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: RA3-6B2]

Lieferant: Tonbo Biosciences
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

50-0452-U100 50-0452-U025
50-0452-U100EA 57.6 EUR
50-0452-U100 50-0452-U025
Anti-CD45R Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: RA3-6B2]
Antikörper
The RA3-6B2 antibody reacts with the human and mouse CD45 isoform known as CD45R, or B220, a protein tyrosine phosphatase of 220 kDa. CD45 is one of the most abundant hematopoietic markers, and is expressed on all leukocytes (the Leukocyte Common Antigen, LCA). Various isoforms are generated and expressed in cell-specific patterns, all critical for leukocyte function. In mouse, the CD45R/B220 isoform is predominantly found on B cells, at varying levels on all stages from pro-B cells to activated B cells, and may also be detected on certain T cell and NK cell subsets. It is of note that B220 is not similarly expressed on human B cells, where it appears to be differentiation-specific and therefore expressed on only some B cell subsets. Other forms of CD45 with restricted cellular expression include CD45RA, CD45RB and CD45RO.

The RA3-6B2 antibody is one of the most consistently used leukocyte markers for B cells, T cell subsets and NK cell subsets in human and mouse. This antibody has also been reported as cross-reactive with feline CD45R/B220.

Type: Primary
Antigen: CD45R (CD45R)
Clonality: monoclonal
Clone: RA3-6B2
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a Kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR