Drucken
Centrifuge, refrigerated, KR4i
Lieferant: JOUAN
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
11178160
521-0389EA 0 EUR
521-0389
Centrifuge, refrigerated, KR4i
Zentrifugen Standzentrifugen
This high-capacity, low-speed centrifuge accepts large sample numbers and volumes and delivers outstanding performance.
  • Blood banking capability allows processing of up to 12 quin or quad blood bags with or without filters
  • Guaranteed lifetime reproducibility, run to run by automatic High Reproducibility Factor (integral) - Ideal for platelet production
  • Careful sample handling: Liners that enable blood bags to be inserted and removed without force
  • Temperature sensor and software independent of the bowl for accurate sample temperature control
  • Intuitive VIDEOset™ graphic microprocessor control system with pictures and message
  • Automatic rotor recognition prevents rotor overspeed
Ideal for high-throughput clinical applications and genomics, proteomics and biochemistry research.