Drucken
Static ionizer, ION-100A
Lieferant: OHAUS
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
30095929
611-3318EA 1510 EUR
611-3318
Static ionizer, ION-100A
Ionisatoren
Electrostatic charges on weighing containers and samples can result in false readings or instability as they create attractive and repulsive forces. The ION-100A is designed to be used in conjunction with an OHAUS balance to neutralise electrostatic charges that can lead to inaccuracies.
  • Increases the accuracy of lab measurements
  • Fan-free technology neutralises static with no sample disruption
  • Compact and versatile design; small footprint
  • Durable electrodes can be easily replaced
The ionizer neutralises the electrostatic charge between the test tube and the balance, increasing the accuracy of measurements - particularly with low-volume samples.