Drucken
Industrial balances, Defender™ 3000 washdown
Lieferant: OHAUS
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
80252934UK 80252933UK 80252932UK 80252931UK 80252930UK 80252929UK 30685138
OHAU80252934UKEA 0 EUR
OHAU80252934UK OHAU80252933UK OHAU80252932UK OHAU80252931UK OHAU80252930UK OHAU80252929UK 611-5195
Industrial balances, Defender™ 3000 washdown
Waagen
A range of modular scales for tough industrial environments. Easy to use scales that are ideal for general weighing and simple counting applications in production, packaging, and shipping and receiving areas.
  • IP 66 stainless steel indicator
  • Sturdy tubular platform design with high quality 304 stainless steel top plate
  • Internal rechargeable battery