Drucken
Injection and aspiration filter, Sterifix® Pury
Lieferant: B. Braun
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
4551001
612-0192EA 154 EUR
612-0192
Injection and aspiration filter, Sterifix® Pury
Infusionen
Injection and aspiration filter for the prevention of particulate contamination during injection and aspiration of drugs.