Drucken
Blood collection tubes, BD Microtainer®
Lieferant: BECTON DICKINSON MEDICAL
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
365975 365954 365993 365986 365988 365968 365964 365966
613-3969EA 452 EUR
613-3969 BDAM365954 BDAM365993 BDAM365986 BDAM365988 BDAM365968 BDAM365964 BDAM365966
Blood collection tubes, BD Microtainer®
Klinisches Diagnosezubehör Klinische Probenentnahme und Probenvorbereitung Entnahme und Vorbereitung von Blutproben
These tubes are used for the collection of capillary blood from skin puncture.
  • Tubes come with a variety of additives for various tests
  • Provided with visible fill lines and are all colour-coded for ease identification
  • Tubes are also available for light sensitive analytes
BD Microtainer® tubes with BD Microgard™ closure have an integrated collection scoop, along with a wider diameter (8 mm) for ease of collection and improved mixing ensuring a quality sample is collected.
Additionally closure helps protect the healthcare worker from exposure toMore Product Information