Drucken
Pipette tips for Rainin® LTS style pipettes, xTIP®
Lieferant: BIOTIX
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
R-0250-9RC
613-5454EA 62.5 EUR
613-5454
Pipette tips for Rainin® LTS style pipettes, xTIP®
Pipettenspitzen
FlexFit®, X-Resin®, and Blade® technologies developed by Biotix are found exclusively in Biotix and xTIP brand pipette tips. Biotix technologies minimise inaccuracies associated with the number one issue in pipetting: user error. These proven features are standard on all xTIP low retention pipette tips, maximising accuracy and precision.
  • Perfect fit thanks to FlexFit technology: FlexFit’s flexible proximal end ensures confidence in a secure seal
  • Clean delivery with X-Resin and Blade technologies
  • X-Resin is a sleek resin that needs no coating, resulting in uniform composition throughout xTIP from proximal end to distal end
  • X-Resin provides ultra-low retentive qualities, with every lot, from the first tip to the last
  • Blade technology eliminates hanging droplet formation, ensuring sample precision with each and every delivery by reducing surface tension at the distal end of the tip
  • Certified free of RNase, DNase, pyrogens, PCR inhibitors
TipEject™ reload system offers convenience and eco-friendliness. Designed for intuitive use and engineered for simple application, TipEject™ saves time, effort, and waste. Quality sterile tips are stored in translucent packaging, fully protected from exposure to environment and handling. Made from sturdy polymer construction, TipEject™ is 100% recyclable. Included in each TipEject™ package are 10 trays (96 tips/tray) and a rack base.
Zertifizierungen: Manufactured under stringent controls in an ISO 9001 certified facility. Each and every lot is tested for precision, ensuring superior quality and compatibility. All raw materials are traceable.
Verpackung: Each tray includes a label with a unique bar-coded serial number, part number, and description for specific traceability.