Drucken
Active gauge displays
Lieferant: EDWARDS HIGH VACUUM METTLER
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
D39565000 D39566000 D39567000 D39585000 D39561000
619-0223EA 0 EUR
619-0223 619-0224 619-0225 EDWAD39585000 EDWAD39561000
Active gauge displays
Pumpen Vakuum-Messgeräte
The active gauge displays are a range of simple, low cost units for the measurement and control of vacuum from atmosphere to 10-9 mbar.
  • Single switch with LED indication for gauge head selection
  • Option of two adjustable set-points with relay status indication
  • Mains power input supply via IEC connector
  • Clear analog display covers the whole gauge head pressure range
  • Available with both bench top use or panel mounting
Lieferumfang: Supplied with tilt feet for convenient bench top mounting and panel mounting kit to DIN 43700 standard.