Drucken
Mica microhub, Leica®
Lieferant: LEICA MICROSYSTEMS
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
11889183_P1
630-3484EA 0 EUR
630-3484
Mica microhub, Leica®
Mikroskope
Mica - The world's first imaging Microhub. More than a highly automated microscope, Mica unites widefield and confocal imaging in a sample protecting, incubating environment. With the simple push of a button, you have everything you need - all in one place - to supercharge fluorescence imaging workflows and get meaningful scientific results faster.
  • Focus on your science, not figuring out your microscope
  • Access for all
  • No constraints
  • Radically simplified workflows
Intelligent automation - All opto-digital components are fully motorised andMore Product Information
Order Now
[{"stiboOrderingTable":"<!--Specifications Tab-->\r\n\t\t\t\t<h2>TECHNISCHE DATEN</h2>\r\n\t\t\t\t <div id="stiboSpecGrid">\r\n\t\t\t\t\t<table id="stiboSpecGridTable" width="100%" class="productGrid">\r\n\t\t\t\t\t\t<caption class="caption">\r\n\t\t\t\t\t\t</caption>\r\n\t\t\t\t\t\t<tbody>\r\n\t\t\t\t\t\t\t<tr>\r\n\t\t\t\t\t\t\t\t<th></th>\t\t\t\t\t\t\t\t\t\t\t\r\n\t\t\t\t\t\t\t</tr>\r\n\t\t\t\t\t\t\t<tr class="productGrid">\t\r\n\t\t\t<td class="productGrid bld" colspan="1">Environmentally Preferable</td>\r\n\t\t\t<td class="productGrid bld" colspan="1"></td>\r\n\t\t\t</tr>\r\n\t\t<tr class="productGrid">\t\r\n\t\t\t<td class="productGrid bld" colspan="1">Beobachtungsmethode</td>\r\n\t\t\t<td class="productGrid bld" colspan="1">Hellfeld und Fluoreszenz</td>\r\n\t\t\t</tr>\r\n\t\t<style type="text/css">\r\n \t.stibospecprodgrid {\r\n \t\t\tdisplay:block;\r\n \t\t\toverflow-x:auto;\r\n \t\t\t}\t\r\n</style>\r\n\r\n<script type="text/javascript">\r\n\r\n\t$(document).ready(function() {\r\n\t\tif (numCol("#stiboSpecGridTable") > 4){\r\n\t\t\t$('#stiboSpecGrid').addClass('stibospecprodgrid');\r\n\t\t}\t\t\r\n\t});\r\n\t\r\n\tfunction numCol(table) {\r\n\t var maxColNum = 0;\r\n\t var i=0;\r\n\t var trs = $(table).find("tr");\r\n\t for ( i=0; i<trs.length; i++ ) {\r\n\t maxColNum = Math.max(maxColNum, getColForTr(trs[i]));\r\n\t }\r\n\r\n\t return maxColNum;\r\n\t}\r\n\tfunction getColForTr(tr) {\r\n\r\n\t var tds = $(tr).find("td");\r\n\r\n\t var numCols = 0;\r\n\r\n\t var i=1;\r\n\t for ( i=1; i<tds.length; i++ ) {\r\n\t var span = $(tds[i]).attr("colspan");\r\n\r\n\t if ( span )\r\n\t numCols += parseInt(span);\r\n\t else {\r\n\t numCols++;\r\n\t }\r\n\t }\r\n\t return numCols;\r\n\t}\r\n</script>\t\t\t\t\t\t\t\t\r\n</tbody>\r\n\t\t\t\t\t</table>\r\n\t\t\t\t\t</div>"}]