Drucken
Flexacam C5 and I5 Microscope Cameras
Lieferant: LEICA MICROSYSTEMS
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
12730537 12730535 12730536
630-3528EA 1790 EUR
630-3528 630-3526 630-3529
Flexacam C5 and I5 Microscope Cameras
Kameras
With Flexacam microscope cameras, you can see the fine sample details you need to see with precise colors and high resolution.
  • Flexacam c5 microscope camera for C-mount installation on a stereo or compound microscope
  • Flexacam i5 microscope camera for installation between the optics carrier and binocular tube of a stereo or compound microscope
  • Reveal fine details of your sample and capture images in high resolution thanks to the 12-megapixel CMOS sensor
  • Differentiate details quickly and clearly due to the fast auto exposure, even when switching between bright and dark samples
  • Visualise sample movements clearly and smoothly on the monitor, facilitated by the high frame rate of up to 60 fps
With the Flexacam c5 and i5 microscope cameras, you can efficiently reveal sample insights taking advantage of sharp images showing fine details with precise colours. Additionally, you can work in a flexible way according to your requirements and make team decisions more easily using either camera option.
The microscope cameras can be operated with the consistent user interface of the Enersight software platform, no matter the operation mode, i.e., a computer, monitor, or mobile device.
The cameras are powered by the Enersight software platform, so that you can enjoy an easy-to-use and consistent interface.
Zertifizierungen: DoC CE.
Lieferumfang: Flexacam c5 includes HDMI cable, USB C-A cable, external power supply (without power cord), USB stick.
Flexacam i5 (compound) includes HDMI cable, USB C-A cable, USB stick.
Flexacam i5 (stereo) includes HDMI cable, USB C-A cable, external power supply (without power cord), USB stick.