Drucken
Contact/Non contact tachometer
Lieferant: TQC
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
LU2000
649-0037EA 269.44 EUR
649-0037
Contact/Non contact tachometer
Tachometer
This digital tachometer is designed to be used for both contact and non-contact measurements.
  • Memory for the maximum, minimum and last measured values
  • Can also be used to measure a wheel speed
  • High visible display with low battery indicator
Lieferumfang: Supplied in a carrying case with reflecting tape marks (700 mm) and manual.