Drucken

Anti-CD45 Mouse Antibody (PerCP-Cy5.5) [clone: HI30]

Lieferant: Tonbo Biosciences
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

65-0459-T100
65-0459-T100EA 213 EUR
65-0459-T100
Anti-CD45 Mouse Antibody (PerCP-Cy5.5) [clone: HI30]
Antikörper
The HI30 antibody reacts with human CD45, one of the most abundant hematopoietic markers and one that is expressed on all leukocytes (the Leukocyte Common Antigen, LCA). CD45 is a protein tyrosine phosphatase existing in several isoforms, each being generated and expressed in cell-specific patterns. With its broad cell distribution, CD45 is critical for many leukocyte functions, regulating signal transduction and cell activation associated with the T cell receptor, B cell receptor, and IL-2 receptor. Other forms of CD45, with restricted cellular expression, include CD45R (B220), CD45RA, CD45RB, CD45RO and others.

The HI30 antibody is widely used as a marker for human CD45 expression on T cells, B cells, monocytes, macrophages, and NK cells.

Type: Primary
Antigen: CD45
Clonality:
Clone: HI30
Conjugation: PerCP-Cy5.5
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR