Drucken

pH buffer solutions, NIST traceable, Orion™

Lieferant: THERMO ORION
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

910760 910460 910199 910105 910112 910168 9104CB 910686 9107CB 910918 911060 9110CB 911260-WA 9116860 916860 9191860 910104 910107 910110 916099
ORIO910760EA 86.5 EUR
ORIO910760 ORIO910460 662-1549 662-1551 662-1550 ORIO910168 ORIO9104CB ORIO910686 ORIO9107CB 662-0978 ORIO911060 ORIO9110CB ORIO911260-WA ORIO9116860 ORIO916860 ORIO9191860 662-0770 662-0771 662-0772 ORIO916099
pH buffer solutions, NIST traceable, Orion™
Puffer pH-Puffer

Perform accurate, reproducible pH calibrations with Orion™ pH buffers.


  • NIST traceability
  • Colour-coded solutions are available for ease of use and identification


Solutions are manufactured under ISO 9000 quality standards.


The kit includes 1 pint (475 ml) bottle each of pH 4,00 buffer, pH 7,00 buffer, pH 10,01 buffer and standard pH electrode storage solution and one pH electrode storage bottle.


Lieferumfang: Supplied in 475 ml or 60 ml bottles, with buffer/temperature tables to ensure more precise calibration.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR