Drucken
pH/mV/°C meter, handheld, Eutech pH 150
Lieferant: EUTECH
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
ECPHWP15003K ECPHWP15000 ECPHWP15002 ECPHWP15002K
662-2311EA 651 EUR
662-2311 662-2300 662-2272 662-2273
pH/mV/°C meter, handheld, Eutech pH 150
Elektrochemische Messgeräte pH- und Multi-Parameter-Messgeräte
This portable meter features a waterproof design with IP67 rating.
  • Large, three-line LCD displays pH or mV values
  • Grip-Clip™: Quickly and easily secures the pH electrode to a beaker or container
  • Automatic buffer recognition; up to 5- or 6-point calibration; calibration due alarm
  • Stores up to 150 measurements along with temperature
  • Can be used with two AA batteries or with optional universal power adapter (not included)
  • Built-in stand with wall-mount option allows for easy bench top measurement or storage
The meter can be used with pH electrodes with BNC connector.
Lieferumfang: Meter only (without electrodes) is supplied with Grip-Clip™ and two AA batteries.
Set 1 additionally includes a double junction pH electrode and ATC probe.
Set 2 additionally includes a double-junction pH electrode and ATC probe, hard carry case, pH buffers, storage solution and rinse solution.
Set 3 additionally includes a double-junction pH/ATC electrode, hard carry case, pH buffers, storage solution and rinse solution.