Drucken
Conductivity meter, handheld, Eutech CON 150
Lieferant: EUTECH
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
ECCONWP15003 ECCONWP15003K
663-0343EA 513 EUR
663-0343 663-0344
Conductivity meter, handheld, Eutech CON 150
Elektrochemische Messgeräte Leitfähigkeitsmessgeräte
This portable meter features a waterproof design with IP67 rating.
  • Large, three line LCD displays conductivity or TDS measurement with temperature as well as cell efficiency
  • Grip-Clip™: Quickly and easily secures the pH electrode to a beaker or container
  • Stores up to 150 measurements along with temperature
  • Adjustable temperature coefficient – Set to 0,0 % to meet USP methods for non compensated measurements
  • Calibrate using automatically recognised standards or manually; calibration due alarm
  • Can be used with two AA batteries or with optional universal power adapter (not included)
  • Built-in stand with wall-mount option allows for easy bench top measurement or storage
The meter accepts probes with a 0,1; 1,0 or 10 cell constant.
Lieferumfang: Meter is supplied with conductivity cell (K = 1.0), Grip-Clip™ and two AA batteries.
The kit includes meter, conductivity cell (K = 1.0), Grip-Clip™, two AA batteries, hard carrying case and calibration solution.