Drucken
Conductivity meter, handheld, Eutech COND 6+
Lieferant: EUTECH
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
ECCON603PLUS ECCON603PLUSK
663-0350EA 478 EUR
663-0350 663-0353
Conductivity meter, handheld, Eutech COND 6+
Elektrochemische Messgeräte Leitfähigkeitsmessgeräte
This portable economic meter with large LCD display is ideal for quick, accurate conductivity checks in laboratories, field and schools.
  • Option of quick, easy automatic calibration or customised, near-to-sample manual calibration
  • Auto-ranging for conductivity measurements for fast response and best resolution over wide measurement range
  • HOLD function and auto-power off
  • Self-diagnostic with message codes for easy troubleshooting
  • Electrode with built-in ATC designed for minimal air bubble entrapment during measurement
  • Built with shock-resistant ABS plastic and protected by an IP54-rated housing against accidental splashes
  • Protective rubber boot shields meter from drops and features a sturdy built-in stand for easy bench top operation
Useful in nutrient and fertiliser checks in hydroponics and agricultural industries. Also suitable for analysing water hard water, untreated water, industrial and rinse water, drinking water, effluent water, pool water and incoming process water. Ideal for all types of quality assurance, printing industries and water quality testing.
Lieferumfang: Supplied with conductivity electrode ECCONSEN91B.
The kit additionally includes the conductivity carrying kit (includes 1× 1413 uS, 12,88 mS, 84 uS KCl standard solution and 1× deionised (rinse) water (60 ml each).