Thrombin
Lieferant: Novagen (Part of Merck)Gesamtbewertung: 0
Bewertung: von 5
Restriction grade thrombin is a highly purified preparation qualified to specifically cleave target proteins produced with appropriate vectors. This preparation is functionally tested for activity with fusion proteins and is free of detectable contaminating proteases.
Prepared from human plasma that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV. Thrombin is supplied as a solution in 50% glycerol and includes 10X Thrombin cleavage buffer and a cleavage control protein.