Drucken
Anti-PTPRC Rat Monoclonal Antibody [clone: 30-F11]
Lieferant: Tonbo Biosciences
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
70-0451-U100
70-0451-U100EA 67.1 EUR
70-0451-U100
Anti-PTPRC Rat Monoclonal Antibody [clone: 30-F11]
Antikörper
The 30-F11 antibody reacts with mouse CD45, which is one of the most abundant hematopoietic markers and is expressed on all leukocytes (the Leukocyte Common Antigen, LCA). CD45 is a protein tyrosine phosphatase existing in several isoforms, each being generated and expressed in cell-specific patterns. With its broad cell distribution, CD45 is critical for many leukocyte functions, regulating signal transduction and cell activation associated with the T cell receptor, B cell receptor, and IL-2 receptor. Other forms of CD45, with restricted cellular expression, include CD45R (B220), CD45RA, CD45RB and CD45RO.
The 30-F11 antibody is widely used as a leukocyte marker for B cells, T cell subsets and NK cell subsets.
Type: Primary
Antigen: PTPRC (protein tyrosine phosphatase, receptor type, C)
Clonality: monoclonal
Clone: 30-F11
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2b Kappa
Reactivity: Mouse