Drucken
Combs for gel casting system, MultiCaster
Lieferant: EMBITEC
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
EP-1026 EP-1027 EP-1028
730-0361EA 96.2 EUR
730-0361 730-0362 730-0363
Combs for gel casting system, MultiCaster
Elektrophorese-Systeme Horizontale Gelelektrophoresesysteme
Reversible combs for the MultiCaster gel casting system with dual configuration for multiple gel formats.
  • Made of polycarbonate
  • Well configurations and estimated sample volume: 12+1 well (30 μl) and 24+1 well (11 μl)
  • Multi-channel pipette compatibility: 12-channel
  • Estimated sample volume is based on gel thickness of 6,5 mm
All combs are interchangeable so that a single MultiCaster system can be used for all preparations and high throughput needs by simply swapping the combs.