Drucken
Visualisierungsplatte
Lieferant: EMBITEC
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
EP-1047 EP-1048
730-2202EA 14.3 EUR
730-2202 730-2203
Visualisierungsplatte
Elektrophorese-Systeme Horizontale Gelelektrophoresesysteme
RunOne™ well visualisation plates are designed to make gel loading simple. The light colour of most gel units provides little or no contrast for the agarose gel and the gel wells. RunOne™ well visualisation plates are red, fit securely into the RunOne™ system, and provide a stable platform for gel running.