Drucken

Guanidiniumchlorid ≥99,5%, GenAR® für die Biotechnologie, Macron Fine Chemicals™

Lieferant: MACRON AVANTOR BRAND
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

Achtung

7716-87EA 0 EUR
7716-87 7716-88
Guanidiniumchlorid ≥99,5%, GenAR® für die Biotechnologie, Macron Fine Chemicals™
Guanidiniumchlorid

Tested for absence of detectable aldehydes and ammeline. DNase-, RNase-, and protease-free.

Formel: NH₂C(=NH)NH₂·HCl
MW: 95.53 g/mol
Melting Pt: 185 °C
Dichte: 1.354 g/cm³ (20 °C)
Storage Temperature: Raumtemperatur
MDL Number: MFCD00013026
CAS-Nummer: 50-01-1
EINECS: 200-002-3
Merck Index: 13,04578
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da5a871cab977e6d9dc666b457732eb1e869a714&vid=ec49804319d0557e26e13f7611afdfc1d7f091c3&prid=web&clid=SAPDAM

Order Now

Spezifikation Testergebnisse

For Laboratory,Research,or Manufacturing Use = 99.5 %
Assay (NH₂C(:NH)NH₂·HCl) ≥99.5 %
Acidity (as HCl) ≤0.01 %
Copper (Cu) ≤10 ppm
Heavy Metals (as Pb) ≤10 ppm
Iron (Fe) ≤10 ppm
Loss on Drying at 105°C ≤0.5 %
Enzyme Activity - DNase Activity Passes Test
Enzyme Activity - RNase Activity Passes Test
Enzyme Activity - Protease Activity Passes Test
Absorbance of a 6M Solution (260 nm) (au) ≤0.030
Residue after Ignition ≤0.20 %
MANUFACTURED EXCLUSIVELY FOR AVANTOR PERFORMANCE MATERIALS, LLC., BY
BIOSPECTRA, INC., STROUDSBURG, PA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR