Periodic table mix 3 for ICP, 16 elements, TraceCERT®, Supelco®
Lieferant: SUPELCO
Ratings:
(keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
78334-100ML
78334-100MLEA
275
EUR
78334-100ML
Periodic table mix 3 for ICP, 16 elements, TraceCERT®, Supelco®
Standards
Kationischer Standards
Mehrelement-Standards
10 mg/l each: Sc, Y, La, Ce, Pr, Nd, Sm, Eu, Gd, Tb, Dy, Ho, Er, Tm, Yb and Lu in 5% nitric acid
Zertifizierungen: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034