Drucken
Blocking buffers, FirstChoice™
Lieferant: G Bioscienes
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
786-666 786-667 786-668 786-669
786-666EA 246 EUR
786-666 786-667 786-668 786-669
Blocking buffers, FirstChoice™
Puffer Elektrophorese-Puffer Blockier-Puffer
FirstChoice™ Blocking Buffer is ideal as a first choice for optimization of new assays and systems, or when determining the optimal blocking buffer for elimination of non specific binding sites in ELISA, blotting, immunohistochemistry and other applications. Buffers are conveniently supplied in widely used TBS (Tris-buffered saline at pH 7,5) and PBS (phosphate-buffered saline at pH 7,5) buffers as well as in separate formulations containing Tween® 20 for improving blocking efficiencies.
  • Ready to use
  • Greater versatility
  • Lack of cross reactivity
  • Compatible with antibodies and avidin/biotin based system