Immobilised Pepstatin for purification of Cathepsins/Pepsin/Pepstatin binding molecules
Lieferant: G Bioscienes
Ratings:
(keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
786-789
786-789EA
807
EUR
786-789
Immobilised Pepstatin for purification of Cathepsins/Pepsin/Pepstatin binding molecules
Protein-Aufbereitungs
Protein Purification Resins
Immobilised Pepstatin is for the purification of cathepsins, pepsin, bacterial aspartic proteases, HIV proteases and other molecules that bind pepstatin.