Triethylcitrat 99,0-101,0 alkalimetry on anhydrous substance NF, Ph. Eur., JPE, SAFC®
Lieferant: MERCK PRODUCTION CHEMICALS
Ratings:
(keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
8.17059.1000
8.17059.9025
8.17059.1000EA
412
EUR
8.17059.1000
8.17059.9025
Triethylcitrat 99,0-101,0 alkalimetry on anhydrous substance NF, Ph. Eur., JPE, SAFC®
Triethylcitrat
Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).
Zertifizierungen: Corresponds to Ph. Eur., JPE, NF
Formel:
C₁₂H₂₀O₇ MW: 276.29 g/mol Siedepunkt: 294 Melting Pt: –55 °C Dichte: 1.135 g/cm³ (25 °C) Flash-Pt: 150 Storage Temperature: Raumtemperatur |
MDL Number:
MFCD00009201 CAS-Nummer: 77-93-0 EINECS: 201-070-7 |
Spezifikation Testergebnisse
Assay (alkalimetric, calculated on anhydrous substance) | 99.0 - 101.0 % |
Assay (GC, calc. on anhydrous substance) | 99.0 - 100.5 % |
Identity (IR-spectrum) | passes test |
Identity (GC) | passes test |
Identity (wet chemistry) | passes test |
Identity (Refractive index) | passes test |
Appearance | clear solution, not more intense in color than matching fluid A (1:10) and reference solution BY₆ |
Acidity | passes test |
Refractive index (n 20/D) | 1.440 - 1.446 |
Refractive index (n 25/D) | 1.439 - 1.441 |
Density (d 25/25) | 1.135 - 1.139 |
Density (d 20/20) | 1.135 - 1.145 |
Viscosity (20 °C) | 26 - 38 mPa∙s |
Sulfate (SO₄) | ≤ 150 ppm |
Heavy metals (as Pb) | ≤ 5 ppm |
As (Arsenic) | ≤ 2 ppm |
Cyclohexane (HS-GC) | ≤ 3880 ppm |
Ethanol (HS-GC) | ≤ 5000 ppm |
Other residual solvents (ICH (Q3C)) | excluded by manufacturing process |
Related substances (GC) (largest single impurity) | ≤ 0.2 % |
Related substances (GC) (Sum of all impurities) | ≤ 0.5 % |
Triethyl aconitate (GC) | ≤ 0.2 % |
Related substances (GC) | passes test |
Sulfated ash (600 °C) | ≤ 0.1 % |
Water (according to Karl Fischer) | ≤ 0.25 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...