Drucken
Tape dispenser, multiple roll
Lieferant: ULTRATAPE
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
UT924
817-0310EA 800 EUR
817-0310
Tape dispenser, multiple roll
Klebeband-Abroller
This multiple roll dispenser is designed to hold four rolls of up to 25,4 mm width and 201,2 metres in length.
Lieferumfang: Supplied with four 76 mm plastic core holders.