Ferrocen 99
Lieferant: ALFA AESARGesamtbewertung: 0
Bewertung: von 5
|
Gefahr
|
Argon_filled
Purity: 99%
CAS No.: 102-54-5
Molecular Formula: C10H10Fe
Formula Weight: 186.04
Physical Form: Powder, Packaged under argon
Sensitivity: Air and Moisture Sensitive
Appearance: Yellow to orange
Odour: Characteristic
Melting Point: 173-174°
Boiling Point: 249°
Merck Reference: 14,4037
UN No.: 1325
MDL No.: MFCD00001427
Formel:
C₁₀H₁₀Fe MW: 186.04 g/mol Siedepunkt: 249 °C Melting Pt: 173…174 °C Dichte: 1.49 g/cm³ (20 °C) Storage Temperature: Raumtemperatur |
MDL Number:
MFCD00001427 CAS-Nummer: 102-54-5 EINECS: 203-039-3 UN: 1325 ADR: 4.1 II Merck Index: 14,4037 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...