Colour Reference Solutions acc. to USP, analytical standards, Supelco®
Lieferant: SUPELCOGesamtbewertung: 0
Bewertung: von 5
These are sets of the following colour reference solutions according to USP:
Colour Reference Solution A
Colour Reference Solution B
Colour Reference Solution C
Colour Reference Solution D
Colour Reference Solution E
Colour Reference Solution F
Colour Reference Solution G
Colour Reference Solution H
Colour Reference Solution I
Colour Reference Solution J
Colour Reference Solution K
Colour Reference Solution L
Colour Reference Solution M
Colour Reference Solution N
Colour Reference Solution O
Colour Reference Solution P
Colour Reference Solution Q
Colour Reference Solution R
Colour Reference Solution S
Colour Reference Solution T
Bestellinformation: Available in 2 ml and 10 ml ampoules.
Achtung! Bitte beachten: Contains Cobalt(II) chloride. Please refer to SDS for hazards.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...