Drucken

Acetonitril, ultralow_water 99,5 GC, corrected for water content, BakerDRY™, J.T.Baker®

Lieferant: AVANTOR PERFORMANCE MATERIALS
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

Gefahr

9035-12EA 277.48 EUR
9035-12 9035-10
Acetonitril, ultralow_water 99,5 GC, corrected for water content, BakerDRY™, J.T.Baker®
Acetonitril

Specifically manufactured for use in organic, organometallic, and oligonucleotide synthesis requiring low water solvents.

Formel: H₃CCN
MW: 41.05 g/mol
Siedepunkt: 81.6
Melting Pt: –45.7 °C
Dichte: 0.782 g/cm³ (20 °C)
Flash-Pt: 2 °C (geschlossener Tiegel)
Storage Temperature: Raumtemperatur
MDL Number: MFCD00001878
CAS-Nummer: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3 II
REACH: 01-2119471307-38
Merck Index: 13,00071
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b965ef40b6e61dd29b33363d25c3df2038fea764&prid=web&clid=SAPDAM

Order Now

Spezifikation Testergebnisse

For Laboratory,Research,or Manufacturing Use
ACS - Assay (CH₃CN) (by GC, corrected for water) ≥99.5 %
Appearance Passes Test
ACS - Color (APHA) ≤10
ACS - Residue after Evaporation ≤1 ppm
ACS - Titrable Acid (µeq/g) ≤0.8
ACS - Titrable Base (µeq/g) ≤0.6
ACS - Water (by KF, coulometric) ≤10.0 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR