Transition metal mix 3 for ICP, 7 elements, TraceCERT®, Supelco®
Lieferant: SUPELCO
Ratings:
(keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
93468-100ML
93468-100MLEA
311
EUR
93468-100ML
Transition metal mix 3 for ICP, 7 elements, TraceCERT®, Supelco®
Standards
Kationischer Standards
Mehrelement-Standards
100 mg/l each: Ru, Rh, Pd, Os, Ir, Pt and Au in 10% hydrochloric acid
Zertifizierungen: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034