Drucken

N-Boc-glycin (Boc-Gly-OH) ≥98%

Lieferant: ALFA AESAR
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

Gefahr

A11579.06 A11579.14 A11579.22
A11579.06EA 26.2 EUR
A11579.06 A11579.14 A11579.22
N-Boc-glycin (Boc-Gly-OH) ≥98%
N-Boc-glycin (Boc-Gly-OH)

Purity: 98+% CAS No.: 4530-20-5 Molecular Formula: (CH3)3COCONHCH2CO2H Formula Weight: 175.18 Melting Point: 87-90° Beilstein Reference: 1101514

Formel: C₇H₁₃NO₄
MW: 175.2 g/mol
Melting Pt: 87…90 °C
Storage Temperature: Raumtemperatur
MDL Number: MFCD00002690
CAS-Nummer: 4530-20-5
EINECS: 224-864-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR