p-Anisaldehyd 98
Lieferant: ALFA AESARGesamtbewertung: 0
Bewertung: von 5
|
Achtung
|
Purity: 98%
CAS No.: 123-11-5
Molecular Formula: C8H8O2
Formula Weight: 136.15
Flash Point: 116°(241°F)
Physical Form: Liquid
Density: 1.121
Refractive Index: 1.5730
Sensitivity: Air and Light Sensitive
Appearance: Colourless to pale yellow
Odour: Odourless
Melting Point: 0-2°
Boiling Point: 247-249°
Beilstein Reference: 471382
Merck Reference: 14,663
MDL No.: MFCD00003385
Formel:
C₈H₈O₂ MW: 136.15 g/mol Siedepunkt: 247…249 °C Melting Pt: 0…2 °C Dichte: 1.121 Flash-Pt: 116 °C (241 °F) Storage Temperature: Raumtemperatur |
MDL Number:
MFCD00003385 CAS-Nummer: 123-11-5 EINECS: 204-602-6 Merck Index: 13,00666 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...