Drucken

2-(Cyclohexylamino)ethansulfonsäure (CHES) ≥99%

Lieferant: ALFA AESAR
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

Achtung

A18047.14 A18047.22 A18047.36 A18047.99
A18047.14EA 47.1 EUR
A18047.14 A18047.22 A18047.36 A18047.99
2-(Cyclohexylamino)ethansulfonsäure (CHES) ≥99%
2-(Cyclohexylamino)ethansulfonsäure (CHES)

Purity: 99% CAS No.: 103-47-9 Molecular Formula: C8H17NO3S Formula Weight: 207.29 Melting Point: 300° Beilstein Reference: 2967601 Merck Reference: 14,2055 MDL No.: MFCD00003835


Zwitterionic buffer useful in pH range 8.6 to 10.0

Formel: C₈H₁₇NO₃S
MW: 207.29 g/mol
Melting Pt: >300 °C
Storage Temperature: Raumtemperatur
MDL Number: MFCD00003835
CAS-Nummer: 103-47-9
EINECS: 203-115-6
Merck Index: 13,02063

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR