Drucken

Anti-SLC7A11 Rabbit Polyclonal Antibody

Lieferant: Abcam
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

AB37185-100
ABCAAB37185-100EA 980 EUR
ABCAAB37185-100
Anti-SLC7A11 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SLC7A11 (solute carrier family 7 (anionic amino acid transporter light chain, xc- system), member 11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR