Drucken

Anti-XPA Mouse Monoclonal Antibody [clone: 5F12]

Lieferant: Abcam
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

AB65963-100
ABCAAB65963-100EA 939 EUR
ABCAAB65963-100
Anti-XPA Mouse Monoclonal Antibody [clone: 5F12]
Antikörper
Type: Primary
Antigen: XPA (xeroderma pigmentosum, complementation group A)
Clonality: monoclonal
Clone: 5F12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR