Drucken

Anti-Laminin beta 1 Rabbit Polyclonal Antibody

Lieferant: Abcam
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

AB65986-100
ABCAAB65986-100EA 932 EUR
ABCAAB65986-100
Anti-Laminin beta 1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Laminin beta 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR