Drucken

Anti-SOCS1 Rabbit Polyclonal Antibody

Lieferant: Abcam
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

AB65989-100
ABCAAB65989-100EA 918 EUR
ABCAAB65989-100
Anti-SOCS1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SOCS1 (suppressor of cytokine signaling 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR