Drucken

Anti-EFCAB6 Mouse polyclonal antibody unconjugated

Lieferant: Abcam
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

AB68748-50
ABCAAB68748-50EA 871 EUR
ABCAAB68748-50
Anti-EFCAB6 Mouse polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: EFCAB6 (EF-hand calcium binding domain 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR