Drucken

Anti-DERL3 Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

AB78233-50
ABCAAB78233-50EA 932 EUR
ABCAAB78233-50
Anti-DERL3 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: DERL3 (derlin 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR