Drucken

Anti-LMX1A Rabbit Polyclonal Antibody

Lieferant: Abcam
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

AB82833-100
ABCAAB82833-100EA 918 EUR
ABCAAB82833-100
Anti-LMX1A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LMX1A (LIM homeobox transcription factor 1, alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR